4-Brom-2-thiophencarbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Bromo-2-thiophenecarboxaldehyde, 4-Bromothiophene-2-carboxaldehyde

M-Clarity™ Qualitätsstufe = MQ200

283452-25G 283452-5G
SIAL283452-25GEA 247 EUR
SIAL283452-25G SIAL283452-5G
4-Brom-2-thiophencarbaldehyd, Sigma-Aldrich®
4-Brom-2-thiophencarbaldehyd
Formel: C₅H₃BrOS
Molecular Weight: 191.05 g/mol
MDL: MFCD00005431
CAS-Nummer: 18791-75-8
EINECS: 242-577-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR