Perfluor(methylcyclohexan), Sigma-Aldrich®

Lieferant: Merck

Synonyme: FlutecR PP2

M-Clarity™ Qualitätsstufe = MQ100

302937-100G 302937-25G
SIAL302937-100GEA 164 EUR
SIAL302937-100G SIAL302937-25G
Perfluor(methylcyclohexan), Sigma-Aldrich®
Perfluor(methylcyclohexan)
Formel: C₇F₁₄
Molecular Weight: 350.05 g/mol
MDL: MFCD00001468
CAS-Nummer: 355-02-2
EINECS: 206-573-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR