3-Chlorphenethylamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(3-Chlorophenyl)ethylamine
M-Clarity™ Qualitätsstufe = MQ100
340847-25G
340847-5G
SIAL340847-25GEA
186
EUR
SIAL340847-25G
SIAL340847-5G
3-Chlorphenethylamin, Sigma-Aldrich®
3-Chlorphenethylamin
Formel:
C₈H₁₀ClN Molecular Weight: 155,63 g/mol |
MDL:
MFCD00047957 CAS-Nummer: 13078-79-0 EINECS: 235-982-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...