Tetramethylammoniumhydrogensulfat Hydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

358967-10G 358967-50G
SIAL358967-10GEA 74.4 EUR
SIAL358967-10G SIAL358967-50G
Tetramethylammoniumhydrogensulfat Hydrat, Sigma-Aldrich®
Tetramethylammoniumhydrogensulfat Hydrat
Formel: C₄H₁₃NO₄S·H₂O
Molecular Weight: 189,23 g/mol
CAS-Nummer: 207738-07-6
EINECS: 279-490-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR