2-Fluorphenethylamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Amino-2-(2-fluorophenyl)ethane, 2-Fluorophenethylamine, 2-(2-Fluorophenyl)ethylamine
M-Clarity™ Qualitätsstufe = MQ100
368032-25G
368032-5G
SIAL368032-25GEA
275
EUR
SIAL368032-25G
SIAL368032-5G
2-Fluorphenethylamin, Sigma-Aldrich®
2-Fluorphenethylamin
Formel:
C₈H₁₀FN Molecular Weight: 139,17 g/mol |
MDL:
MFCD00075502 CAS-Nummer: 52721-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...