Acenaphthen-[D10], Sigma-Aldrich®

Lieferant: Merck

Synonyme: Acenaphthene-[²H₁₀]

M-Clarity™ Qualitätsstufe = MQ200

451819-1G 451819-5G
SIAL451819-1GEA 243 EUR
SIAL451819-1G SIAL451819-5G
Acenaphthen-[D10], Sigma-Aldrich®
Acenaphthen-[D10]
Formel: C₁₂D₁₀
Molecular Weight: 164,13 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 15067-26-2
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR