4-(Methylthio)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Methylthio)benzeneboronic acid, 4-Thioanisoleboronic acid

M-Clarity™ Qualitätsstufe = MQ100

456802-5G
SIAL456802-5GEA 66.6 EUR
SIAL456802-5G
4-(Methylthio)phenylboronsäure, Sigma-Aldrich®
4-(Methylthio)phenylboronsäure
Formel: C₇H₉BO₂S
Molecular Weight: 168.02 g/mol
MDL: MFCD00093410
CAS-Nummer: 98546-51-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR