4-Fluor-7-nitrobenzofurazan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Fluoro-7-nitro-[2,1,3]-benzoxadiazole, NBD-F

M-Clarity™ Qualitätsstufe = MQ200

47140-10MG 47140-50MG
SIAL47140-10MGEA 237 EUR
SIAL47140-10MG SIAL47140-50MG
4-Fluor-7-nitrobenzofurazan, Sigma-Aldrich®
4-Fluor-7-nitrobenzofurazan
Formel: C₆H₂FN₃O₃
Molecular Weight: 183,1 g/mol
MDL: MFCD00010196
CAS-Nummer: 29270-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR