Fmoc-Pro-OH, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline

47636-50G-F 47636-5G-F
SIAL47636-50G-FEA 81 EUR
SIAL47636-50G-F SIAL47636-5G-F
Fmoc-Pro-OH, Sigma-Aldrich®
Fmoc-Pro-OH
Formel: C₂₀H₁₉NO₄
Molecular Weight: 337,38 g/mol
MDL: MFCD00037122
CAS-Nummer: 71989-31-6
EINECS: 276-259-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR