2-Benzo[b]thiophenboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Benzo[b]thiophen-2-ylboronic acid, 2-Benzothienylboronic acid
M-Clarity™ Qualitätsstufe = MQ100
499978-25G
499978-5G
SIAL499978-25GEA
432
EUR
SIAL499978-25G
SIAL499978-5G
2-Benzo[b]thiophenboronsäure, Sigma-Aldrich®
2-Benzo[b]thiophenboronsäure
Formel:
C₈H₇BO₂S Molecular Weight: 178.02 g/mol |
MDL:
MFCD01075674 CAS-Nummer: 98437-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...