5-Formyl-2-thiophenboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Formylthiophen-2-boronic acid, 5-Formylthiophene-2-boronic acid
M-Clarity™ Qualitätsstufe = MQ200
514055-1G
514055-5G
SIAL514055-1GEA
42.2
EUR
SIAL514055-1G
SIAL514055-5G
5-Formyl-2-thiophenboronsäure, Sigma-Aldrich®
5-Formyl-2-thiophenboronsäure
Formel:
C₅H₅BO₃S Molecular Weight: 155,97 g/mol |
MDL:
MFCD02093666 CAS-Nummer: 4347-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...