3,4-Dimethylbenzoesäure ≥98%

Lieferant: Thermo Scientific
Achtung

A19260.14 A19260.22 A19260.36
A19260.14EA 45.4 EUR
A19260.14 A19260.22 A19260.36
3,4-Dimethylbenzoesäure ≥98%
3,4-Dimethylbenzoic acid

Purity: 98% CAS No.: 619-04-5 Molecular Formula: C9H10O2 Formula Weight: 150.18 Odour: Odourless Melting Point: 164-167° Beilstein Reference: 907267 MDL No.: MFCD00002524

Formel: C₉H₁₀O₂
Molecular Weight: 150,18 g/mol
Siedepunkt: 295 °C (1013 hPa)
Schmelzpunkt: 164…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002524
CAS-Nummer: 619-04-5
EINECS: 210-576-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR