4-Methylbenzamidoxim, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Hydroxy-4-methylbenzimidamide

M-Clarity™ Qualitätsstufe = MQ100

548707-1G 548707-5G
SIAL548707-1GEA 33.3 EUR
SIAL548707-1G SIAL548707-5G
4-Methylbenzamidoxim, Sigma-Aldrich®
4-Methylbenzamidoxim
Formel: C₈H₁₀N₂O
Molecular Weight: 150,18 g/mol
MDL: MFCD00019952
CAS-Nummer: 19227-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR