3-Bromphenethylbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Bromo-3-(2-bromoethyl)benzene

653802-10G 653802-1G
SIAL653802-10GEA 323 EUR
SIAL653802-10G SIAL653802-1G
3-Bromphenethylbromid, Sigma-Aldrich®
3-Bromphenethylbromid
Formel: C₈H₈Br₂
Molecular Weight: 263,96 g/mol
MDL: MFCD07784359
CAS-Nummer: 40422-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR