2-Fluorphenethylbromid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-(2-Bromoethyl)-2-fluorobenzene
655023-1G
655023-5G
SIAL655023-1GEA
50.8
EUR
SIAL655023-1G
SIAL655023-5G
2-Fluorphenethylbromid, Sigma-Aldrich®
2-Fluorphenethylbromid
Formel:
C₈H₈BrF Molecular Weight: 203,05 g/mol |
MDL:
MFCD04117466 CAS-Nummer: 91319-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...