2-Methoxyphenethylbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2-Bromoethyl)-2-methoxybenzene

655031-1G 655031-5G
SIAL655031-1GEA 86.1 EUR
SIAL655031-1G SIAL655031-5G
2-Methoxyphenethylbromid, Sigma-Aldrich®
2-Methoxyphenethylbromid
Formel: C₉H₁₁BrO
Molecular Weight: 215,09 g/mol
MDL: MFCD07369785
CAS-Nummer: 36449-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR