3-Methoxyphenethylbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2-Bromoethyl)-3-methoxybenzene

M-Clarity™ Qualitätsstufe = MQ100

655333-1G 655333-5G
SIAL655333-1GEA 54.1 EUR
SIAL655333-1G SIAL655333-5G
3-Methoxyphenethylbromid, Sigma-Aldrich®
3-Methoxyphenethylbromid
Formel: C₉H₁₁BrO
Molecular Weight: 215,09 g/mol
MDL: MFCD07369789
CAS-Nummer: 2146-61-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR