4'-Fluor-3'-nitroacetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

676357-1G 676357-5G
SIAL676357-1GEA 37.2 EUR
SIAL676357-1G SIAL676357-5G
4'-Fluor-3'-nitroacetophenon, Sigma-Aldrich®
4'-Fluor-3'-nitroacetophenon
Formel: C₈H₆FNO₃
Molecular Weight: 183,14 g/mol
MDL: MFCD00115369
CAS-Nummer: 400-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR