Tetramethylammoniumhydrogensulfat, Supelco®

Lieferant: Merck

Synonyme: Tetramethylammonium hydrogensulphate, Tetramethylammonium bisulphate

M-Clarity™ Qualitätsstufe = MQ100

87724-10G-F 87724-50G-F
87724-10G-FEA 357 EUR
87724-10G-F 87724-50G-F
Tetramethylammoniumhydrogensulfat, Supelco®
Tetramethylammonium hydrogen sulphate
Formel: C₄H₁₃NO₄S
MDL: MFCD00036149
CAS-Nummer: 80526-82-5
EINECS: 279-490-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4651ae55de3053434799889a9abbcd8f4784d91d&vid=e48768f22880db16368900698207f25ca9c6cb9e&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR