8-Amino-2-naphthalinsulfonsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Naphthylamine-7-sulphonic acid, 1,7-Cleve's acid
M-Clarity™ Qualitätsstufe = MQ100
A7782-100G
A7782-25G
SIALA7782-100GEA
203
EUR
SIALA7782-100G
SIALA7782-25G
8-Amino-2-naphthalinsulfonsäure, Sigma-Aldrich®
8-Amino-2-naphthalenesulphonic acid
Formel:
C₁₀H₉NO₃S Molecular Weight: 223.25 g/mol |
MDL:
MFCD00004032 CAS-Nummer: 119-28-8 EINECS: 204-311-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...