N-(3-Aminopropyl)morpholin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-(4-Morpholinyl)propylamine, 3-Morpholinopropan-1-amine, 3-Morpholino propylamine
A9028-100ML
A9028-500ML
SIALA9028-100MLEA
47.6
EUR
SIALA9028-100ML
SIALA9028-500ML
N-(3-Aminopropyl)morpholin, Sigma-Aldrich®
N-(3-Aminopropyl)morpholin
Formel:
C₇H₁₆N₂O Molecular Weight: 144,22 g/mol |
MDL:
MFCD00006184 CAS-Nummer: 123-00-2 EINECS: 204-590-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...