Benzyltrimethylammoniumhydroxid 40% in Methanol, Sigma-Aldrich®
Lieferant: Merck
Synonyme: Triton B, Benzyltrimethylammonium Hydroxide, N,N,N-Trimethyl-1-phenylmethanaminium Hydroxide
M-Clarity™ Qualitätsstufe = MQ200
B32602-500ML B32602-100ML
SIALB32602-500MLEA 263 EUR
SIALB32602-500ML SIALB32602-100ML
Benzyltrimethylammoniumhydroxid 40% in Methanol, Sigma-Aldrich®
Benzyltrimethylammoniumhydroxid
Formel: C₁₀H₁₇NO
Molecular Weight: 167.25
MDL: MFCD00008281
CAS-Nummer: 100-85-6
EINECS: 202-895-5
UN: 2924
ADR: 3 II