Benzyldimethylhexadecylammoniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Cetalkonium chloride, N-Benzyl-N,N-dimethylhexadecan-1-aminium chloride

M-Clarity™ Qualitätsstufe = MQ200

B4136-25G B4136-100G
SIALB4136-25GEA 74.6 EUR
SIALB4136-25G SIALB4136-100G
Benzyldimethylhexadecylammoniumchlorid, Sigma-Aldrich®
Benzyldimethylhexadecylammoniumchlorid
Formel: C₂₅H₄₆ClN
Molecular Weight: 396,10 g/mol
MDL: MFCD00011625
CAS-Nummer: 122-18-9
EINECS: 204-526-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR