2-Azepanon, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Azepan-2-one, 2-Oxohexamethyleneimine, 6-Aminocaproic acid lactam

M-Clarity™ Qualitätsstufe = MQ200

C2204-1KG C2204-250G C2204-5G
SIALC2204-1KGEA 45.9 EUR
SIALC2204-1KG SIALC2204-250G SIALC2204-5G
2-Azepanon, Sigma-Aldrich®
ε-Caprolactam
Formel: C₆H₁₁NO
Molecular Weight: 113,16 g/mol
MDL: MFCD00006936
CAS-Nummer: 105-60-2
EINECS: 203-313-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR