4-Chlorphenethylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(4-Chlorophenyl)ethylamine

M-Clarity™ Qualitätsstufe = MQ100

C65408-25G C65408-5G
SIALC65408-25GEA 92 EUR
SIALC65408-25G SIALC65408-5G
4-Chlorphenethylamin, Sigma-Aldrich®
4-Chlorphenethylamin
Formel: C₈H₁₀ClN
Molecular Weight: 155.63 g/mol
MDL: MFCD00008191
CAS-Nummer: 156-41-2
EINECS: 205-853-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR