Dichlordiphenylsilan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diphenyldichlorosilane, Dichlorodiphenylsilane

M-Clarity™ Qualitätsstufe = MQ100

D61504-100G D61504-5G
SIALD61504-100GEA 95.6 EUR
SIALD61504-100G SIALD61504-5G
Dichlordiphenylsilan, Sigma-Aldrich®
Dichloro(diphenyl)silane
Formel: C₁₂H₁₀Cl₂Si
Molecular Weight: 253,2 g/mol
MDL: MFCD00000489
CAS-Nummer: 80-10-4
EINECS: 201-251-0
UN: 1769
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR