Dichlordiphenylsilan
Lieferant: CHEMPUR SILANE
Synonyme:
Diphenyldichlorosilane, Dichlorodiphenylsilane
SILS07800.25
S07800.100
SILNSILS07800.25EA
0
EUR
SILNSILS07800.25
FLUOS07800.100
Dichlordiphenylsilan
Dichloro(diphenyl)silane
Formel:
C₁₂H₁₀Cl₂Si Molecular Weight: 253,2 g/mol |
MDL:
MFCD00000489 CAS-Nummer: 80-10-4 EINECS: 201-251-0 UN: 1769 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...