1-(Trimethylsiloxy)cyclohexen

Lieferant: CHEMPUR SILANE

Synonyme: (Cyclohex-1-en-1-yloxy)trimethylsilane

SILS18775.10 SILS18775.25 SILS18775.100
SILNSILS18775.10EA 0 EUR
SILNSILS18775.10 SILNSILS18775.25 SILNSILS18775.100
1-(Trimethylsiloxy)cyclohexen
1-(Trimethylsiloxy)cyclohexen
Formel: C₉H₁₈OSi
Molecular Weight: 170.33 g/mol
MDL: MFCD00001541
CAS-Nummer: 6651-36-1
EINECS: 229-675-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR