α-Chlor-o-xylol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Methylbenzyl chloride
A13448.22
A13448.36
A13448.22EA
54.6
EUR
A13448.22
A13448.36
α-Chlor-o-xylol ≥98%
α-Chlor-o-xylol
Purity: 98+%
CAS No.: 552-45-4
Molecular Formula: C8H9Cl
Formula Weight: 140.61
Storage Temperature: 2°C to 8°C
Flash Point: 73°(163°F)
Refractive Index: 1.5410
Odour: Pungent
Melting Point: 2°
Boiling Point: 197-199°
Beilstein Reference: 386016
Merck Reference: 14,10091
UN No.: 3265
MDL No.: MFCD00000899
Formel:
C₈H₉Cl Molecular Weight: 140,61 g/mol Siedepunkt: 197…199 °C Schmelzpunkt: 2 °C Dichte: 1,081 Flammpunkt: 73 °C (163 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00000899 CAS-Nummer: 552-45-4 EINECS: 209-013-8 UN: 3265 ADR: 8,II Merck Index: 14,10091 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...