2-Chlor-m-xylol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Chlor-2,6-dimethylbenzol, 2-Chlor-1,3-dimethylbenzol
A17867.14
A17867.22
A17867.36
A17867.14EA
64.5
EUR
A17867.14
A17867.22
A17867.36
2-Chlor-m-xylol ≥98%
2-Chlor-m-xylol
Purity: 98%
CAS No.: 6781-98-2
Molecular Formula: C8H9Cl
Formula Weight: 140.61
Flash Point: 63°(145°F)
Refractive Index: 1.5270
Melting Point: -36°
Boiling Point: 182-183°
Beilstein Reference: 2040954
MDL No.: MFCD00000565
Formel:
C₈H₉Cl Molecular Weight: 140,61 g/mol Siedepunkt: 182…183 °C Schmelzpunkt: –36 °C Dichte: 1,061 Flammpunkt: 63 °C (145 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000565 CAS-Nummer: 6781-98-2 EINECS: 229-843-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...