α'-Chlor-α,α,α-trifluor-m-xylol

Lieferant: FLUOROCHEM

007609-25G 007609-100G
FLUO007609-25GEA 101 EUR
FLUO007609-25G FLUO007609-100G
α'-Chlor-α,α,α-trifluor-m-xylol
α'-Chlor-α,α,α-trifluor-m-xylol
Formel: C₈H₆ClF₃
Siedepunkt: 95 °C (1013 hPa)
Dichte: 1,3079 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000908
CAS-Nummer: 705-29-3
EINECS: 211-884-4
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR