3-Fluor-o-xylol
Lieferant: FLUOROCHEM
Synonyme:
1,2-Dimethyl-3-fluorbenzol
002577-25G
002577-100G
FLUO002577-25GEA
70.1
EUR
FLUO002577-25G
FLUO002577-100G
3-Fluor-o-xylol
3-Fluor-o-xylol
Formel:
C₈H₉F Molecular Weight: 124,16 g/mol Siedepunkt: 146…148 °C (1013 hPa) Dichte: 0,99 g/cm³ (20 °C) |
MDL:
MFCD00000323 CAS-Nummer: 443-82-3 EINECS: 207-140-3 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...