α,α,α,α',α',α'-Hexafluor-m-xylol
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
α,α,α,β,β,β-Hexafluor-m-xylol
SB00520.1
SB00520.10
SB00520.50
MAYBSB00520.1EA
0
EUR
MAYBSB00520.1
MAYBSB00520.10
MAYBSB00520.50
α,α,α,α',α',α'-Hexafluor-m-xylol
α,α,α,α',α',α'-Hexafluor-m-xylol
Formel:
C₈H₄F₆ Molecular Weight: 214,11 g/mol Siedepunkt: 116,3 °C (1013 hPa) Schmelzpunkt: –35 °C Dichte: 1,383 g/cm³ (22 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000392 CAS-Nummer: 402-31-3 EINECS: 206-939-4 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...