α,α,α-Trifluor-p-xylol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(Trifluormethyl)toluol, 1-Methyl-4-(trifluormethyl)benzol, 4-Methylbenzotrifluorid

L11043.03 L11043.06
L11043.03EA 51.5 EUR
L11043.03 L11043.06
α,α,α-Trifluor-p-xylol ≥98%
α,α,α-Trifluor-p-xylol

Purity: 98% CAS No.: 6140-17-6 Molecular Formula: C8H7F3 Formula Weight: 160.14 Flash Point: 27°(80°F) Refractive Index: 1.4260 Boiling Point: 129° Beilstein Reference: 1863596 UN No.: 1993 MDL No.: MFCD00075476

Formel: C₈H₇F₃
Molecular Weight: 160,14 g/mol
Siedepunkt: 129 °C
Dichte: 1,144
Flammpunkt: 27 °C (80 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00075476
CAS-Nummer: 6140-17-6
EINECS: 228-124-2
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR