α,α,α-Trifluor-p-xylol

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 4-(Trifluormethyl)toluol, 1-Methyl-4-(trifluormethyl)benzol, 4-Methylbenzotrifluorid

009277-1G 009277-5G
FLUO009277-1GEA 0 EUR
FLUO009277-1G FLUO009277-5G
α,α,α-Trifluor-p-xylol
α,α,α-Trifluor-p-xylol
Formel: C₈H₇F₃
Molecular Weight: 160.14 g/mol
Siedepunkt: 120
Dichte: 1.14 g/cm³ (20 °C)
MDL: MFCD00075476
CAS-Nummer: 6140-17-6
EINECS: 228-124-2
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR