α,α,α,α',α',α'-Hexafluor-5-nitroxylol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
1-Nitro-3,5-bis(trifluoromethyl)benzene
006031-25G
006031-100G
FLUO006031-25GEA
64.2
EUR
FLUO006031-25G
FLUO006031-100G
α,α,α,α',α',α'-Hexafluor-5-nitroxylol
α,α,α,α',α',α'-Hexafluor-5-nitroxylol
Formel:
C₈H₃F₆NO₂ Molecular Weight: 259,11 g/mol Siedepunkt: 71 °C (1013 hPa) Dichte: 1,53 g/cm³ (20 °C) |
MDL:
MFCD00000384 CAS-Nummer: 328-75-6 EINECS: 206-336-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...