2-Methyl-3-nitrobenzotrifluorid

Lieferant: FLUOROCHEM

006818-5G 006818-25G 006818-1G
FLUO006818-5GEA 0 EUR
FLUO006818-5G FLUO006818-25G FLUO006818-1G
2-Methyl-3-nitrobenzotrifluorid
2-Methyl-3-nitrobenzotrifluorid
Formel: C₈H₆F₃NO₂
Molecular Weight: 205.14 g/mol
Siedepunkt: 35…38 °C (0.04 mmHg)
MDL: MFCD00042322
CAS-Nummer: 6656-49-1
EINECS: 229-688-2
UN: 2810
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR