2-Iod-p-xylol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2-Iodo-1,4-dimethylbenzene, 1,4-Dimethyl-2-iodobenzene

003509-25G 003509-100G
FLUO003509-25GEA 152 EUR
FLUO003509-25G FLUO003509-100G
2-Iod-p-xylol
2-Iod-p-xylol
Formel: C₈H₉I
Molecular Weight: 232,06 g/mol
MDL: MFCD00013708
CAS-Nummer: 1122-42-5
EINECS: 214-347-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR