4-tert-Butylbenzylamin

Lieferant: FLUOROCHEM
Achtung

Synonyme: p-tert-Butylbenzylamine

008874-1G 008874-5G
FLUO008874-1GEA 57.9 EUR
FLUO008874-1G FLUO008874-5G
4-tert-Butylbenzylamin
4-tert-Butylbenzylamin
Formel: C₁₁H₁₇N
Molecular Weight: 163,26 g/mol
MDL: MFCD00040754
CAS-Nummer: 39895-55-1
EINECS: 254-681-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR