2,3-Dimethylanisol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Methoxy-2,3-dimethylbenzene

A19992.09 A19992.18
A19992.09EA 52.4 EUR
A19992.09 A19992.18
2,3-Dimethylanisol ≥97%
2,3-Dimethylanisol

Purity: 97% CAS No.: 2944-49-2 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 67°(152°F) Refractive Index: 1.5200 Melting Point: 29° Boiling Point: 195° Beilstein Reference: 1859736 MDL No.: MFCD00008376

Formel: C₉H₁₂O
Molecular Weight: 136,19 g/mol
Siedepunkt: 195 °C
Schmelzpunkt: 29 °C
Dichte: 0,984
Flammpunkt: 67 °C (152 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008376
CAS-Nummer: 2944-49-2
EINECS: 220-948-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR