3,5-Xylenol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
sym.-m-Xylenol, 5-Hydroxy-m-xylol, 3,5-Dimethylphenol, 5-Hydroxy-3-xylol, 1,3-Dimethyl-5-hydroxybenzol
A12236.22
A12236.36
AB116245.0500
A12236.22EA
35.2
EUR
A12236.22
A12236.36
ABCRAB116245.0500
3,5-Xylenol ≥98%
3,5-Xylenol
Purity: 98+%
CAS No.: 108-68-9
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 109°(228°F)
Melting Point: 61-65°
Boiling Point: 222°
Beilstein Reference: 774117
Merck Reference: 14,10082
UN No.: 2261
MDL No.: MFCD00002307
Formel:
C₈H₁₀O Molecular Weight: 122.17 g/mol Siedepunkt: 222 °C Schmelzpunkt: 61…65 °C Dichte: 1.12 Flammpunkt: 109 °C (228 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002307 CAS-Nummer: 108-68-9 EINECS: 203-606-5 UN: 2261 ADR: 6.1 II Merck Index: 12,10215 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...