1,4-Diisopropylbenzol 98%

Lieferant: Thermo Scientific
Achtung

B22871.14 B22871.22 B22871.36
B22871.14EA 34.5 EUR
B22871.14 B22871.22 B22871.36
1,4-Diisopropylbenzol 98%
1,4-Diisopropylbenzol

Purity: 99% CAS No.: 100-18-5 Molecular Formula: C12H18 Formula Weight: 162.27 Flash Point: 76°(170°F) Refractive Index: 1.4890 Odour: Aromatic Melting Point: -17° Boiling Point: 210° Beilstein Reference: 1854739 MDL No.: MFCD00008892

Formel: C₁₂H₁₈
Molecular Weight: 162,27 g/mol
Siedepunkt: 210 °C
Schmelzpunkt: –17 °C
Dichte: 0,857
Flammpunkt: 76 °C (170 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008892
CAS-Nummer: 100-18-5
EINECS: 202-826-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR