o-Ethyltoluol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-Ethyl-2-methylbenzol
L02028.06
L02028.14
L02028.06EA
41.9
EUR
L02028.06
L02028.14
o-Ethyltoluol ≥98%
o-Ethyltoluol
Purity: 98+%
CAS No.: 611-14-3
Molecular Formula: C9H12
Formula Weight: 120.20
Flash Point: 53°(127°F)
Refractive Index: 1.5040
Melting Point: -17°
Boiling Point: 164-165°
Beilstein Reference: 1851237
UN No.: 3295
MDL No.: MFCD00009257
Formel:
C₉H₁₂ Molecular Weight: 120,19 g/mol Siedepunkt: 164…165 °C Schmelzpunkt: –17 °C Dichte: 0,884 Flammpunkt: 53 °C (127 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009257 CAS-Nummer: 611-14-3 EINECS: 210-255-1 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...