2,4,6-Trimethylbenzaldehyd ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Mesitylen-2-carbaldehyd, Mesitylaldehyd, Mesitaldehyd
A10104.18
A10104.04
A10104.09
A10104.18EA
208
EUR
A10104.18
A10104.04
A10104.09
2,4,6-Trimethylbenzaldehyd ≥98%
2,4,6-Trimethylbenzaldehyd
Purity: 98%
CAS No.: 487-68-3
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 106°(222°F)
Refractive Index: 1.5540
Melting Point: 14°
Boiling Point: 237°
Beilstein Reference: 1364114
MDL No.: MFCD00003341
Formel:
C₁₀H₁₂O Molecular Weight: 148,2 g/mol Siedepunkt: 237 °C Schmelzpunkt: 14 °C Dichte: 1,005 Flammpunkt: 106 °C (222 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003341 CAS-Nummer: 487-68-3 EINECS: 207-662-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...