2,4,6-Trimethylbenzoesäure
Lieferant: CHEMPUR
Synonyme:
Mesitylen-2-carbonsäure
DP01930.250
DP01930.1
CHMPDP01930.250EA
0
EUR
CHMPDP01930.250
MAYBDP01930.1
2,4,6-Trimethylbenzoesäure
2,4,6-Trimethylbenzoesäure
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2 g/mol Siedepunkt: 298 °C (1013 hPa) Schmelzpunkt: 151…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002481 CAS-Nummer: 480-63-7 EINECS: 207-553-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...