2,4,6-Trimethylphenol 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
140210100
140210500
ACRO140210100EA
27.4
EUR
ACRO140210100
ACRO140210500
2,4,6-Trimethylphenol 99%
2,4,6-Trimethylphenol
Formel:
C₉H₁₂O Molecular Weight: 136.19 g/mol Siedepunkt: 220 °C (760 mmHg) Schmelzpunkt: 70…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002235 CAS-Nummer: 527-60-6 EINECS: 208-419-2 UN: 2430 ADR: 8 II |
Spezifikation Testergebnisse
Appearance | White to yellow to beige Powder or crystals or needles and/or chunks |
Infrared spectrum | Conforms |
Melting point | 70 to 74 °C |
GC | ≥98.5 % |
Specific optical rotation | -16° to -20° (20 °C, 589 nm) c=1, DMF |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...