3,4,5-Trimethylphenol ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

L02911.03 L02911.06 L02911.14
L02911.03EA 58.3 EUR
L02911.03 L02911.06 L02911.14
3,4,5-Trimethylphenol ≥97%
3,4,5-Trimethylphenol

Purity: 97% CAS No.: 527-54-8 Molecular Formula: C9H12O Formula Weight: 136.20 Odour: Phenol-like Melting Point: 104-108° Beilstein Reference: 1907246 UN No.: 2430 MDL No.: MFCD00002305

Formel: C₉H₁₂O
Molecular Weight: 136.19 g/mol
Siedepunkt: 249
Schmelzpunkt: 104…108 °C
Dichte: 0.8184 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002305
CAS-Nummer: 527-54-8
EINECS: 208-418-7
UN: 2430
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR