1,3,5-Tri-tert-butylbenzol ≥97%

Lieferant: Thermo Fisher Scientific

L18832.09 L18832.18
L18832.09EA 79.8 EUR
L18832.09 L18832.18
1,3,5-Tri-tert-butylbenzol ≥97%
1,3,5-Tri-tert-butylbenzol

Purity: 97+% CAS No.: 1460-02-2 Molecular Formula: C18H30 Formula Weight: 246.44 Melting Point: 69-73° Boiling Point: 121-122°/12mm MDL No.: MFCD00008831

Formel: C₁₈H₃₀
Molecular Weight: 246.44 g/mol
Siedepunkt: 121…122 °C (12 mmHg)
Schmelzpunkt: 69…73 °C
Dichte: 2.637 g/cm³ (22 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008831
CAS-Nummer: 1460-02-2
EINECS: 215-952-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR