2,4,6-Tris(brommethyl)mesitylen

Lieferant: Thermo Scientific

Synonyme: 1,3,5-Tris(bromomethyl)-2,4,6-trimethylbenzene, 2,4,6-Tris(bromomethyl)mesitylene

TL00798.1 TL00798.10 TL00798.50
MAYBTL00798.1EA 0 EUR
MAYBTL00798.1 MAYBTL00798.10 MAYBTL00798.50
2,4,6-Tris(brommethyl)mesitylen
2,4,6-Tris(brommethyl)mesitylen
Formel: C₁₂H₁₅Br₃
Molecular Weight: 398,96 g/mol
MDL: MFCD00192554
CAS-Nummer: 21988-87-4
EINECS: 244-697-1
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR