Hexakis(brommethyl)benzol ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1,2,3,4,5,6-Hexakis(bromomethyl)benzene

B22016.03 B22016.06
B22016.03EA 37.1 EUR
B22016.03 B22016.06
Hexakis(brommethyl)benzol ≥98%
Hexakis(brommethyl)benzol

Purity: 98% CAS No.: 3095-73-6 Molecular Formula: C12H12Br6 Formula Weight: 635.65 Melting Point: ca 292° dec. UN No.: 3261 MDL No.: MFCD00182539

Formel: C₁₂H₁₂Br₆
Molecular Weight: 635.65 g/mol
Schmelzpunkt: ∼ 292 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00182539
CAS-Nummer: 3095-73-6
EINECS: 221-443-8
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR